siRNA Details
Gene Symbol: 1700034H15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030669.1 | 5 | 2901 | |||
GenBank mRNA | AK137370.1 | - | 6 | 2739 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700034H15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030669.1 | 5 | 2901 | |||
GenBank mRNA | AK137370.1 | - | 6 | 2739 |