siRNA Details
Gene Symbol: Miat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003718.2 | 5 | 746 | |||
NR_033657.1 | 5 | 790 | ||||
GenBank mRNA | AB300594.1 | - | 5 | 733 | ||
AK038726.1 | - | 5 | 791 | |||
AK053540.1 | - | 5 | 734 |
