siRNA Details
Gene Symbol: Gm7904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003372.1 | 1 | 724 | |||
GenBank mRNA | BC028798.1 | - | 1 | 724 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003372.1 | 1 | 724 | |||
GenBank mRNA | BC028798.1 | - | 1 | 724 |