siRNA Details
Gene Symbol: Rny1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004419.1 | 1 | 41 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U84681.1 | - | 1 | 24 | Chr. - - [-] on Build |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: U84683.
Gene Ontology Categories:
