siRNA Details
Gene Symbol: D130009I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015593.2 | 2 | 572 | |||
GenBank mRNA | AK051169.1 | - | 2 | 576 | ||
AK051495.1 | - | 2 | 572 | |||
AK051771.1 | - | 2 | 577 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D130009I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015593.2 | 2 | 572 | |||
GenBank mRNA | AK051169.1 | - | 2 | 576 | ||
AK051495.1 | - | 2 | 572 | |||
AK051771.1 | - | 2 | 577 |