siRNA Details
Gene Symbol: D130009I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015593.2 | 5 | 1576 | |||
GenBank mRNA | AK051495.1 | - | 5 | 1576 |
Gene Symbol: Gm20597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037973.1 | 4 | 1617 | |||
GenBank mRNA | AK035871.1 | - | 4 | 1618 |
