siRNA Details
Gene Symbol: Gm10069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028592.1 | 1 | 598 | |||
NR_028593.1 | 1 | 598 | ||||
GenBank mRNA | AK029950.1 | - | 1 | 300 | ||
AK046088.1 | - | 1 | 554 | |||
AK086937.1 | - | 1 | 599 | |||
BC091771.1 | - | 1 | 16 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG970567.1 | - | 1 | 16 | Chr. 6 - 128439339 - 128503258 [+] on Build GRCm38 |
