siRNA Details
Gene Symbol: Rnf138rt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006700.1 | - | 1 | 37 | ||
BY238719.1 | - | 1 | 38 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706692.1 | - | 1 | 37 | Chr. X - 163760478 - 163761329 [-] on Build GRCm38 |
