siRNA Details
Gene Symbol: Fam120aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015601.1 | 2 | 603 | |||
GenBank mRNA | AK014080.1 | - | 2 | 333 | ||
AK021143.1 | - | 2 | 295 | |||
BC024672.1 | - | 428 | ||||
BC045201.1 | - | 2 | 535 |
