siRNA Details
Gene Symbol: Mir763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030434.1 | 1 | 110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ418834.1 | - | 1 | 449 | Chr. 10 - 120447961 - 120448411 [-] on Build GRCm38 |
