siRNA Details
Gene Symbol: Mir760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030439.1 | 1 | 59 |
Gene Symbol: LOC100047044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147615.1 | - | 1 | 77 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NR_032295.
