siRNA Details
Gene Symbol: 1700086L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030733.1 | 4 | 558 | |||
NR_030734.1 | 4 | 553 | ||||
NR_030735.1 | 3 | 331 | ||||
GenBank mRNA | AK005679.1 | - | 2 | 226 | ||
AK007020.1 | - | 3 | 327 | |||
BG083361.2 | - | 4 | 540 |
