siRNA Details
Gene Symbol: 1700022A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003953.1 | 2 | 342 | |||
GenBank mRNA | AK006230.1 | - | 2 | 342 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706316.1 | - | 2 | 342 | Chr. 5 - 24645453 - 24648338 [+] on Build GRCm38 |
