siRNA Details
Gene Symbol: A930003A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015488.1 | 1 | 91 | |||
NR_027896.1 | 1 | 91 | ||||
GenBank mRNA | AK086168.1 | - | 1 | 92 | ||
AK153879.1 | - | 1 | 93 | |||
BC053981.1 | - | 1 | 37 | |||
BY101678.1 | - | 1 | 92 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX518163.1 | - | 1 | 68 | Chr. 16 - 19877043 - 19884250 [-] on Build GRCm38 |
