siRNA Details
Gene Symbol: A930003A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015488.1 | 1 | 310 | |||
NR_027896.1 | 1 | 310 | ||||
GenBank mRNA | AK086168.1 | - | 1 | 311 | ||
AK153879.1 | - | 1 | 312 | |||
BC053981.1 | - | 1 | 256 | |||
BY101678.1 | - | 1 | 311 |
