siRNA Details
Gene Symbol: A930003A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015488.1 | 2 | 947 | |||
NR_027896.1 | 3 | 841 | ||||
GenBank mRNA | AK020802.1 | - | 2 | 571 | ||
AK020919.1 | - | 2 | 601 | |||
AK086168.1 | - | 3 | 842 | |||
AK153879.1 | - | 2 | 949 | |||
BC053981.1 | - | 2 | 893 |
