siRNA Details
Gene Symbol: AI427809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033139.1 | 2 | 236 | |||
NR_033140.1 | 2 | 326 | ||||
GenBank mRNA | AK144542.1 | - | 2 | 237 | ||
AK171862.1 | - | 2 | 255 | |||
BC086687.1 | - | 2 | 326 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CO428746.1 | - | 2 | 326 | Chr. 4 - 53262799 - 53265492 [-] on Build GRCm38 |
