siRNA Details
Gene Symbol: A330040F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015503.1 | 2 | 1006 | |||
GenBank mRNA | AK052621.1 | - | 2 | 1006 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB497135.2 | - | 2 | 588 | Chr. 19 - 12585870 - 12596148 [-] on Build GRCm38 |
