siRNA Details
Gene Symbol: Firre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081581.1 | - | 1 | 559 | ||
BC066100.1 | - | 18 | 3193 | |||
KJ638686.1 | - | 7 | 1533 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF714932.1 | - | 1 | 103 | Chr. X - 50567581 - 50569745 [-] on Build GRCm38 |
