siRNA Details
Gene Symbol: Firre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081581.1 | - | 1 | 162 | ||
BC066100.1 | - | 18 | 2797 | |||
KJ638686.1 | - | 7 | 1137 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Firre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081581.1 | - | 1 | 162 | ||
BC066100.1 | - | 18 | 2797 | |||
KJ638686.1 | - | 7 | 1137 |