siRNA Details
Gene Symbol: Firre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030946.1 | - | 2 | 371 | ||
BC066100.1 | - | 2 | 176 | |||
BU704013.1 | - | 2 | 362 | |||
KJ638684.1 | - | 2 | 135 | |||
KJ638685.1 | - | 2 | 135 | |||
KJ638686.1 | - | 2 | 330 |
