siRNA Details
Gene Symbol: 1700120K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027915.1 | 1 | 306 | |||
GenBank mRNA | AK007016.1 | - | 1 | 299 | ||
AK007155.1 | - | 1 | 299 | |||
AK018960.1 | - | 1 | 307 | |||
AK018969.1 | - | 1 | 306 | |||
BC049550.1 | - | 291 |
