siRNA Details
Gene Symbol: 2310068J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028124.1 | 1 | 262 | |||
GenBank mRNA | AK010113.1 | - | 1 | 264 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV449410.1 | - | 1 | 212 | Chr. 15 - 99972816 - 99973236 [-] on Build GRCm38 |
