siRNA Details
Gene Symbol: 9230102K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028438.1 | 1 | 421 | |||
GenBank mRNA | AK078991.1 | - | 1 | 423 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA277195.1 | - | 1 | 206 | Chr. 10 - 110600876 - 110615375 [+] on Build GRCm38 |
