siRNA Details
Gene Symbol: 1700108J01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015532.1 | 2 | 255 | |||
GenBank mRNA | AK006443.1 | - | 2 | 225 | ||
AK018942.1 | - | 2 | 256 | |||
AK018945.1 | - | 2 | 226 | |||
AK018955.1 | - | 2 | 226 | |||
BY707218.1 | - | 2 | 256 |
