siRNA Details
Gene Symbol: BC052688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028430.1 | 4 | 912 | |||
GenBank mRNA | AK167253.1 | - | 4 | 909 | ||
BC052688.1 | - | 4 | 868 |
Gene Symbol: Gm5665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC032201.1 | - | 1503 |
