siRNA Details
Gene Symbol: AA387883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030678.1 | 2 | 551 | |||
GenBank mRNA | AK043996.1 | - | 2 | 552 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ917305.1 | - | 2 | 556 | Chr. 19 - 52923194 - 52926129 [+] on Build GRCm38 |
