siRNA Details
Gene Symbol: C630043F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027923.1 | 1 | 193 | |||
GenBank mRNA | AK012513.1 | - | 1 | 117 | ||
AK039121.1 | - | 1 | 125 | |||
AK041409.1 | - | 1 | 195 | |||
AK042990.1 | - | 93 | ||||
AK043880.1 | - | 1 | 184 | |||
AK053841.1 | - | 1 | 157 | |||
AK083285.1 | - | 1 | 114 | |||
AK083963.1 | - | 1 | 129 |
