siRNA Details
Gene Symbol: C630043F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027923.1 | 1 | 844 | |||
GenBank mRNA | AK039121.1 | - | 1 | 776 | ||
AK041409.1 | - | 1 | 846 | |||
AK042990.1 | - | 744 | ||||
AK043880.1 | - | 1 | 836 | |||
AK053841.1 | - | 1 | 808 | |||
AK083285.1 | - | 1 | 764 | |||
AK083963.1 | - | 1 | 780 |
