siRNA Details
Gene Symbol: C630043F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027923.1 | 1 | 1020 | |||
GenBank mRNA | AK039121.1 | - | 1 | 952 | ||
AK041409.1 | - | 1 | 1022 | |||
AK043880.1 | - | 1 | 1012 | |||
AK053841.1 | - | 1 | 984 | |||
AK083285.1 | - | 1 | 940 | |||
AK083963.1 | - | 1 | 956 |
