siRNA Details
Gene Symbol: C630043F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027923.1 | 1 | 1316 | |||
GenBank mRNA | AK039121.1 | - | 1 | 1248 | ||
AK041409.1 | - | 1 | 1318 | |||
AK043880.1 | - | 1 | 1309 | |||
AK053841.1 | - | 1 | 1280 | |||
AK083285.1 | - | 1 | 1237 | |||
AK083963.1 | - | 1 | 1252 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB431992.2 | - | 1 | 549 | Chr. 4 - 72202034 - 72202665 [+] on Build GRCm38 |
