siRNA Details
Gene Symbol: C630043F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027923.1 | 1 | 1775 | |||
GenBank mRNA | AK041409.1 | - | 1 | 1777 | ||
AK043880.1 | - | 1 | 1768 | |||
AK053841.1 | - | 1 | 1739 |
