siRNA Details
Gene Symbol: BC018473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003364.1 | 2 | 457 | |||
GenBank mRNA | AF118272.1 | - | 2 | 406 | ||
AF118273.1 | - | 2 | 455 | |||
AF118847.1 | - | 2 | 382 | |||
AK156340.1 | - | 2 | 295 | |||
BC018473.1 | - | 1 | 644 | |||
BY753224.1 | - | 2 | 300 |
