siRNA Details
Gene Symbol: BC018473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003364.1 | 4 | 857 | |||
GenBank mRNA | AF118272.1 | - | 4 | 806 | ||
AF118273.1 | - | 5 | 966 | |||
AF118847.1 | - | 4 | 782 | |||
AK156340.1 | - | 4 | 696 | |||
BC018473.1 | - | 3 | 1044 |
