siRNA Details
Gene Symbol: BC018473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003364.1 | 4 | 1042 | |||
GenBank mRNA | AF118272.1 | - | 4 | 991 | ||
AF118273.1 | - | 5 | 1151 | |||
AF118558.1 | - | 1 | 131 | |||
AF118847.1 | - | 4 | 967 | |||
AK156340.1 | - | 4 | 882 | |||
BC018473.1 | - | 3 | 1229 |
