siRNA Details
Gene Symbol: 2610035D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015556.1 | 3 | 789 | |||
GenBank mRNA | AK011682.1 | - | 3 | 332 | ||
AK031204.1 | - | 3 | 790 | |||
BC024713.1 | - | 2 | 728 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610035D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015556.1 | 3 | 789 | |||
GenBank mRNA | AK011682.1 | - | 3 | 332 | ||
AK031204.1 | - | 3 | 790 | |||
BC024713.1 | - | 2 | 728 |