siRNA Details
Gene Symbol: 1700026D11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028286.1 | 3 | 466 | |||
GenBank mRNA | AK006376.1 | - | 3 | 467 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX519837.1 | - | 4 | 355 | Chr. 2 - 132490751 - 132523858 [+] on Build GRCm38 |
