siRNA Details
Gene Symbol: A330023F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK171382.1 | - | 1409 | |||
BC058715.1 | - | LIT3442 C030002C11Rik |
1 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB723210.1 | - | 1 | 79 | Chr. 1 - 195024109 - 195034161 [+] on Build GRCm38 |
