siRNA Details
Gene Symbol: A330023F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | BC058715.1 | - | LIT3442 C030002C11Rik |
3 | 3946 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A330023F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | BC058715.1 | - | LIT3442 C030002C11Rik |
3 | 3946 |