siRNA Details
Gene Symbol: A330023F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015566.2 | 8 | 3783 | |||
GenBank mRNA | AK045637.1 | - | 8 | 3784 | ||
BC058715.1 | - | LIT3442 C030002C11Rik |
3 | 3024 |
