siRNA Details
Gene Symbol: 4933439C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015585.2 | 3 | 1395 | |||
GenBank mRNA | AK011602.1 | - | 3 | 928 | ||
AK017115.1 | - | 3 | 1396 | |||
AK030920.1 | - | 1423 | ||||
AK051100.1 | - | 2 | 931 | |||
AK163429.1 | - | 3 | 1391 | |||
AK169756.1 | - | 3 | 1425 | |||
BC065402.1 | - | 2 | 101 | |||
BC099591.1 | - | 3 | 642 |
