siRNA Details
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028086.1 | 2 | 2383 | |||
GenBank mRNA | AK050814.1 | - | 2 | 2149 | ||
AK078069.1 | - | 2 | 2410 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028086.1 | 2 | 2383 | |||
GenBank mRNA | AK050814.1 | - | 2 | 2149 | ||
AK078069.1 | - | 2 | 2410 |