siRNA Details
Gene Symbol: Smim10l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028086.1 | 2 | 1506 | |||
GenBank mRNA | AK050814.1 | - | 2 | 1272 | ||
AK078069.1 | - | 2 | 1529 | |||
AK139449.1 | - | 2 | 1485 | |||
BC052359.1 | - | 2 | 1477 | 6585408 |
