siRNA Details
Gene Symbol: B430319G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029474.1 | 1 | 2593 | |||
GenBank mRNA | AK044362.1 | - | 1 | 2076 | ||
AK086371.1 | - | 1 | 2500 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B430319G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029474.1 | 1 | 2593 | |||
GenBank mRNA | AK044362.1 | - | 1 | 2076 | ||
AK086371.1 | - | 1 | 2500 |