siRNA Details
Gene Symbol: Pantr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027826.1 | 2 | 207 | |||
GenBank mRNA | AK011437.1 | - | 2 | 207 | ||
AK019154.1 | - | 2 | 207 | |||
BC049671.1 | - | 2 | 212 | |||
BC058417.1 | - | 2 | 300 | |||
BC059892.1 | - | 2 | 281 | |||
BC094545.1 | - | 187 |
