siRNA Details
Gene Symbol: Pantr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027826.1 | 3, 4 | 286 | |||
GenBank mRNA | AK011437.1 | - | 3, 4 | 286 | ||
AK019154.1 | - | 3, 4 | 286 | |||
BC058417.1 | - | 3, 4 | 379 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_183252.1 | - | 286 | Chr. - - [-] on Build |
