siRNA Details
Gene Symbol: 1700069L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033216.1 | 3 | 424 | |||
GenBank mRNA | AK007096.1 | - | 3 | 425 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707170.1 | - | 3 | 425 | Chr. 5 - 113692422 - 113724772 [-] on Build GRCm38 |
