siRNA Details
Gene Symbol: Btbd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024078.1 | 7 | 1617 | |||
GenBank mRNA | AK020599.1 | - | 2 | 764 | ||
AK141215.1 | - | 2 | 849 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_001033187.1 | - | 849 | Chr. - - [-] on Build |
