siRNA Details
Gene Symbol: Btbd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024078.1 | 7 | 1100 | |||
GenBank mRNA | AK020599.1 | - | 2 | 245 | ||
AK141215.1 | - | 2 | 332 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btbd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024078.1 | 7 | 1100 | |||
GenBank mRNA | AK020599.1 | - | 2 | 245 | ||
AK141215.1 | - | 2 | 332 |