siRNA Details
Gene Symbol: Gm5468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027376.1 | 2 | 1119 | |||
GenBank mRNA | AK139483.1 | - | 2 | 254 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB642134.1 | - | 2 | 248 | Chr. 15 - 25415063 - 25450972 [+] on Build GRCm38 |
